missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Chloro-4-methylphenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61690.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496696

  • £47.81 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Chloro-4-methylphenyl)-3,4-difluorobenzenesulfonamide
317.74
MFCD17630183

Specifications

N-(2-Chloro-4-methylphenyl)-3,4-difluorobenzenesulfonamide
97%
MFCD17630183
Solid
317.74
1g
Videos
SDS