missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Chloro-4-iodophenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59381.03

Code : NEW

Additional Details : CAS Number : 424816-35-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15435678

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Chloro-4-iodophenyl)-3-methylbenzamide
424816-35-3
371.6
MFCD02216285

Specifications

N-(2-Chloro-4-iodophenyl)-3-methylbenzamide
Light Sensitive
97%
MFCD02216285
424816-35-3
Solid
371.6
1g
Videos
SDS