missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Carboxyphenyl)glycine, 97%, Thermo Scientific™

Catalog Number p-3757199
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
612-42-0
C9H9NO4
195.174
MFCD00020445
PJUXPMVQAZLJEX-UHFFFAOYSA-N
n-2-carboxyphenyl glycine, 2-carboxymethyl amino benzoic acid, n-carboxymethyl anthranilic acid, phenylglycine-o-carboxylic acid, benzoic acid, 2-carboxymethyl amino, 2-carboxymethyl-amino-benzoic acid, glycine, n-o-carboxyphenyl, 2-carboxymethylamino benzoic acid, anthranilic acid, n-carboxymethyl
69161
2-(carboxymethylamino)benzoic acid
C1=CC=C(C(=C1)C(=O)O)NCC(=O)O
This item is not returnable. View return policy
612-42-0
C9H9NO4
195.174
MFCD00020445
PJUXPMVQAZLJEX-UHFFFAOYSA-N
n-2-carboxyphenyl glycine, 2-carboxymethyl amino benzoic acid, n-carboxymethyl anthranilic acid, phenylglycine-o-carboxylic acid, benzoic acid, 2-carboxymethyl amino, 2-carboxymethyl-amino-benzoic acid, glycine, n-o-carboxyphenyl, 2-carboxymethylamino benzoic acid, anthranilic acid, n-carboxymethyl
69161
2-(carboxymethylamino)benzoic acid
C1=CC=C(C(=C1)C(=O)O)NCC(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 218.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula HO2CC6H4NHCH2CO2H
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 195.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Carboxyphenyl)glycine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'