missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60296.03

Code : NEW

Additional Details : CAS Number : 21226-31-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426768

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromophenyl)benzenesulfonamide
21226-31-3
312.18
MFCD00541883

Specifications

N-(2-Bromophenyl)benzenesulfonamide
1g
97%
312.18
21226-31-3
Solid
White
MFCD00541883
Videos
SDS