missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65175.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429436

  • £85.44 / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
330.23
MFCD28098762

Specifications

N-(2-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
330.23
1g
95%
MFCD28098762
Videos
SDS