missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59497.03

Code : NEW

Additional Details : CAS Number : 36293-14-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15455838

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromophenyl)-3,5-dinitrobenzamide
366.12
2897207
36293-14-8
MFCD00403528

Specifications

N-(2-Bromophenyl)-3,5-dinitrobenzamide
1g
97%
366.12
2897207
36293-14-8
Solid
Off-white to brown
MFCD00403528
Videos
SDS