missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62870.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447936

  • £79.71 / 1g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromophenyl)-3,5-difluorobenzenesulfonamide
348.16
MFCD17629707

Specifications

N-(2-Bromophenyl)-3,5-difluorobenzenesulfonamide
Solid
348.16
1g
97%
MFCD17629707
Videos
SDS