missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4-methylphenyl)-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65354.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429586

  • £56.92 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromo-4-methylphenyl)-4-chloro-3-sulfamoylbenzamide
403.68
MFCD28098788

Specifications

N-(2-Bromo-4-methylphenyl)-4-chloro-3-sulfamoylbenzamide
95%
MFCD28098788
Solid
403.68
1g
Videos
SDS