missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H62913.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide | |
C13H10BrF2NO2S | |
362.19 | |
MFCD25977736 |
Specifications
N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide | |
C13H10BrF2NO2S | |
MFCD25977736 |
97% | |
362.19 | |
5g |