missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62913.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477996

  • £68.26 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide
C13H10BrF2NO2S
362.19
MFCD25977736

Specifications

N-(2-Bromo-4-methylphenyl)-3,5-difluorobenzenesulfonamide
C13H10BrF2NO2S
MFCD25977736
97%
362.19
1g
Videos
SDS