missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4-fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65856.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499946

  • £127.20 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromo-4-fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
348.22
MFCD27975831

Specifications

N-(2-Bromo-4-fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
348.22
Solid
1g
MFCD27975831
Videos
SDS