missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61885.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426836

  • £53.11 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzamide
330.1
MFCD12468855

Specifications

N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzamide
97%
330.1
Solid
1g
MFCD12468855
Videos
SDS