missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4,6-dichlorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H56293.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402606

  • £315.00 / 1g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1421262-63-6
381.065
UYPWKTWHHVRHIG-UHFFFAOYSA-N
73996228
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=C(C=C2Br)Cl)Cl
C12H8BrCl2NO2S
MFCD22682861
n-2-bromo-4,6-dichlorophenyl benzenesulfonamide
N-(2-bromo-4,6-dichlorophenyl)benzenesulfonamide

Specifications

N-(2-Bromo-4,6-dichlorophenyl)benzenesulfonamide
1421262-63-6
MFCD22682861
UYPWKTWHHVRHIG-UHFFFAOYSA-N
N-(2-bromo-4,6-dichlorophenyl)benzenesulfonamide
73996228
97%
1g
C12H8BrCl2NO2S
n-2-bromo-4,6-dichlorophenyl benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=C(C=C2Br)Cl)Cl
381.065
381.07
Videos
SDS Documents
Certificates