missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Aminoethyl)morpholine-4-carboxamide oxalate, 98%

Brand:  Alfa Aesar™ H64743.06

Code : NEW

Additional Details : CAS Number : 154467-16-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419126

  • £29.89 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Aminoethyl)morpholine-4-carboxamide oxalate
154467-16-0
263.25
MFCD11113112

Specifications

N-(2-Aminoethyl)morpholine-4-carboxamide oxalate
154467-16-0
98%
MFCD11113112
5g
Solid
263.25
Videos
SDS