missing translation for 'onlineSavingsMsg'
Learn More

N-2-Acetylguanine, 99%, Thermo Scientific Chemicals

Catalog Number p-3777144
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19962-37-9
C7H7N5O2
193.17
MFCD00078201
MXSMRDDXWJSGMC-UHFFFAOYSA-N
2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide
88319
N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
CC(=O)NC1=NC(=O)C2=C(N1)N=CN2
This item is not returnable. View return policy
19962-37-9
C7H7N5O2
193.17
MFCD00078201
MXSMRDDXWJSGMC-UHFFFAOYSA-N
2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide
88319
N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
CC(=O)NC1=NC(=O)C2=C(N1)N=CN2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point >260°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in acids and alkalies
Formula Weight 193.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-2-Acetylguanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'