Alfa Aesar™ N-(2,5-Dimethylphenyl)-2,2-diphenylacetamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65338.MD

250MG N-(2,5-Dimethylphenyl)-2,2-diphenylacetamide, 95% 250mg

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 70298-67-8 Weight: 0.00025kg


Quantity 250mg

Product Code. 15419576

Quantity Price
1 £ 10.71 / 250mg
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(2,5-Dimethylphenyl)-2,2-diphenylacetamide
CAS 70298-67-8
Formula Weight 315.42
MDL Number MFCD00090603
Beilstein 2145425

Specification

Formula Weight 315.42
Chemical Name or Material N-(2,5-Dimethylphenyl)-2,2-diphenylacetamide
Physical Form Solid
Assay 95%
MDL Number MFCD00090603
CAS 70298-67-8
Beilstein 2145425
Quantity 250mg