missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Difluorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62411.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497406

  • £478.06 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Difluorophenyl)-3,5-difluorobenzenesulfonamide
305.25
MFCD10727857

Specifications

N-(2,5-Difluorophenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD10727857
Solid
305.25
5g
Videos
SDS