missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Difluorophenyl)-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60930.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446116

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Difluorophenyl)-2-nitrobenzenesulfonamide
314.26
MFCD05136526

Specifications

N-(2,5-Difluorophenyl)-2-nitrobenzenesulfonamide
97%
MFCD05136526
Solid
314.26
1g
Videos
SDS