missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Difluorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65509.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432569

  • £74.73 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Difluorophenyl)-2,2-diphenylacetamide
323.34
MFCD04496629

Specifications

N-(2,5-Difluorophenyl)-2,2-diphenylacetamide
95%
MFCD04496629
Solid
323.34
1g
Videos
SDS