missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Diethoxyphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65888.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432659

  • £59.78 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Diethoxyphenyl)-2,2-diphenylacetamide
375.47
MFCD15389849

Specifications

N-(2,5-Diethoxyphenyl)-2,2-diphenylacetamide
95%
MFCD15389849
Solid
375.47
1g
Videos
SDS