missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65288.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489536

  • £79.71 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Dichlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
320.21
MFCD28098778

Specifications

N-(2,5-Dichlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
MFCD28098778
Solid
320.21
1g
Videos
SDS