missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59041.03

Code : NEW

Additional Details : CAS Number : 199726-57-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475218

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Dichlorophenyl)-3-methylbenzamide
199726-57-3
280.15
MFCD00088929

Specifications

N-(2,5-Dichlorophenyl)-3-methylbenzamide
Solid
280.15
1g
199726-57-3
97%
MFCD00088929
Videos
SDS