missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62525.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457526

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Dichlorophenyl)-3,5-difluorobenzenesulfonamide
338.16
MFCD26960677

Specifications

N-(2,5-Dichlorophenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD26960677
Solid
338.16
1g
Videos
SDS