missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-2,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61093.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457588

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Dichlorophenyl)-2,5-difluorobenzamide
302.11
MFCD26143275

Specifications

N-(2,5-Dichlorophenyl)-2,5-difluorobenzamide
97%
MFCD26143275
Solid
302.11
1g
Videos
SDS