missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65308.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412519

  • £109.18 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,5-Dichlorophenyl)-2,2-diphenylacetamide
356.25
MFCD00387788

Specifications

N-(2,5-Dichlorophenyl)-2,2-diphenylacetamide
95%
MFCD00387788
Solid
356.25
1g
Videos
SDS