missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2,5-Bis(trifluoromethyl)phenyl]-3-bromo-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64056.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445655

  • £127.20 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

N-[2,5-Bis(trifluoromethyl)phenyl]-3-bromo-5-(trifluoromethyl)benzenesulfonamide
516.18
MFCD27975662

Specifications

N-[2,5-Bis(trifluoromethyl)phenyl]-3-bromo-5-(trifluoromethyl)benzenesulfonamide
97%
MFCD27975662
Solid
516.18
1g
Videos
SDS