missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2,5-Bis(trifluoromethyl)phenyl]-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62102.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467026

  • £617.98 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-[2,5-Bis(trifluoromethyl)phenyl]-3,5-difluorobenzenesulfonamide
405.26
MFCD26960672

Specifications

N-[2,5-Bis(trifluoromethyl)phenyl]-3,5-difluorobenzenesulfonamide
97%
MFCD26960672
Solid
405.26
5g
Videos
SDS