missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2,5-Bis(trifluoromethyl)phenyl]-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65905.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409996

  • £66.46 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

N-[2,5-Bis(trifluoromethyl)phenyl]-2,2-diphenylacetamide
423.36
MFCD27975840

Specifications

N-[2,5-Bis(trifluoromethyl)phenyl]-2,2-diphenylacetamide
95%
MFCD27975840
Solid
423.36
1g
Videos
SDS