missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dimethoxyphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59133.03

Code : NEW

Additional Details : CAS Number : 328270-05-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415358

  • £358.49 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dimethoxyphenyl)-3-methylbenzamide
328270-05-9
271.32
MFCD00750860

Specifications

N-(2,4-Dimethoxyphenyl)-3-methylbenzamide
Solid
271.32
1g
328270-05-9
97%
MFCD00750860
Videos
SDS