missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Difluorophenyl)thiourea, 97%, Thermo Scientific™

Catalog Number p-7023418
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
175277-76-6
C7H6F2N2S
188.196
MFCD00041150
DZZSKQFBAGZNSH-UHFFFAOYSA-N
2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl, amino 2,4-difluorophenyl amino methane-1-thione, acmc-1bxvl, buttpark 12118-31, difluorophenylthioacetonitrile, 2,4-difluoro-phenyl-thiourea
2734207
(2,4-difluorophenyl)thiourea
C1=CC(=C(C=C1F)F)NC(=S)N
This item is not returnable. View return policy
175277-76-6
C7H6F2N2S
188.196
MFCD00041150
DZZSKQFBAGZNSH-UHFFFAOYSA-N
2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl, amino 2,4-difluorophenyl amino methane-1-thione, acmc-1bxvl, buttpark 12118-31, difluorophenylthioacetonitrile, 2,4-difluoro-phenyl-thiourea
2734207
(2,4-difluorophenyl)thiourea
C1=CC(=C(C=C1F)F)NC(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 165°C to 168°C
Quantity 1 g
UN Number UN2811
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material N-(2,4-Difluorophenyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'