missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Difluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60517.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486988

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Difluorophenyl)benzenesulfonamide
269.27
MFCD01213711

Specifications

N-(2,4-Difluorophenyl)benzenesulfonamide
97%
MFCD01213711
Solid
269.27
1g
Videos
SDS