missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Difluorophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59514.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485858

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Difluorophenyl)-3,5-dinitrobenzamide
323.21
MFCD00751590

Specifications

N-(2,4-Difluorophenyl)-3,5-dinitrobenzamide
97%
MFCD00751590
Solid
323.21
1g
Videos
SDS