missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Difluorophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59942.03

Code : NEW

Additional Details : CAS Number : 314022-67-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466398

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Difluorophenyl)-2-methoxybenzamide
314022-67-8
263.24
MFCD00751585

Specifications

N-(2,4-Difluorophenyl)-2-methoxybenzamide
Solid
263.24
1g
314022-67-8
97%
MFCD00751585
Videos
SDS