missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(2,4-Dichlorophenyl)-2,2-diphenylacetamide, 95%
Brand: Alfa Aesar™ H65753.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(2,4-Dichlorophenyl)-2,2-diphenylacetamide | |
356.25 | |
MFCD00389997 |
Specifications
N-(2,4-Dichlorophenyl)-2,2-diphenylacetamide | |
95% | |
MFCD00389997 |
Solid | |
356.25 | |
1g |