missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-4-chlorobenzamide, 97%

Brand:  Alfa Aesar™ H59813.03

Code : NEW

Additional Details : CAS Number : 541546-46-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486228

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-4-chlorobenzamide
541546-46-7
389.47
MFCD03039734

Specifications

N-(2,4-Dibromophenyl)-4-chlorobenzamide
Solid
389.47
1g
541546-46-7
97%
MFCD03039734
Videos
SDS