missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59174.03

Code : NEW

Additional Details : CAS Number : 551923-55-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465408

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-3-methylbenzamide
551923-55-8
369.06
MFCD01357446

Specifications

N-(2,4-Dibromophenyl)-3-methylbenzamide
Solid
369.06
1g
551923-55-8
97%
MFCD01357446
Videos
SDS