missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62142.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457076

  • £119.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide
427.06
MFCD27975657

Specifications

N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD27975657
Solid
427.06
1g
Videos
SDS