missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62142.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467076

  • £597.84 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide
427.06
MFCD27975657

Specifications

N-(2,4-Dibromophenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD27975657
Solid
427.06
5g
Videos
SDS