missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59607.03

Code : NEW

Additional Details : CAS Number : 304882-34-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495978

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-2-fluorobenzamide
304882-34-6
373.02
MFCD00439075

Specifications

N-(2,4-Dibromophenyl)-2-fluorobenzamide
Solid
373.02
1g
304882-34-6
97%
MFCD00439075
Videos
SDS