missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4,6-Trichlorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H56988.03

Code : NEW

Additional Details : CAS Number : 351027-75-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15403548

  • £315.00 / 1g

Estimated Shipment: 24-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

351027-75-3
336.611
KVDKXSWOSZISRI-UHFFFAOYSA-N
3325605
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=C(C=C2Cl)Cl)Cl
C12H8Cl3NO2S
MFCD00656383
n-2,4,6-trichlorophenyl benzenesulfonamide, phenylsulfonyl 2,4,6-trichlorophenyl amine
N-(2,4,6-trichlorophenyl)benzenesulfonamide

Specifications

N-(2,4,6-Trichlorophenyl)benzenesulfonamide
351027-75-3
MFCD00656383
n-2,4,6-trichlorophenyl benzenesulfonamide, phenylsulfonyl 2,4,6-trichlorophenyl amine
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=C(C=C2Cl)Cl)Cl
336.611
336.62
1g
C12H8Cl3NO2S
3102989
KVDKXSWOSZISRI-UHFFFAOYSA-N
N-(2,4,6-trichlorophenyl)benzenesulfonamide
3325605
97%
Videos
SDS Documents
Certificates