Learn More
Alfa Aesar™ N-(2,3-Dichlorophenyl)-3-methylbenzamide, 97%
Brand: Alfa Aesar™ H59630.03
Code : NEW
Additional Details : CAS Number : 90665-37-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(2,3-Dichlorophenyl)-3-methylbenzamide | |
90665-37-5 | |
280.15 | |
MFCD00586169 |
Specifications
N-(2,3-Dichlorophenyl)-3-methylbenzamide | |
Solid | |
280.15 | |
1g |
90665-37-5 | |
97% | |
MFCD00586169 |