missing translation for 'onlineSavingsMsg'
Learn More

N-(2,3,5,6-Tetramethylphenylsulfonyl)glycine, 96%, Thermo Scientific™

Catalog Number p-7031608
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
379250-94-9
C12H17NO4S
271.33
MFCD02333806
FAIKALCUIWGIHB-UHFFFAOYSA-N
2,3,5,6-tetramethyl-benzenesulfonylamino-acetic acid, 2,3,5,6-tetramethylphenyl sulfonyl glycine, glycine,n-2,3,5,6-tetramethylphenyl sulfonyl, 2-2,3,5,6-tetramethylbenzenesulfonamido acetic acid, 2,3,5,6-tetramethylbenzenesulfonamido acetic acid, n-2,3,5,6-tetramethylphenylsulfonyl glycine, n-2,3,5,6-tetramethylphenyl sulfonyl glycine, 2-2,3,5,6-tetramethylphenylsulfonamido acetic acid, 2,3,5,6-tetramethylbenzenesulfonamidoacetic acid, 2,3,5,6-tetramethylphenyl sulfonyl amino acetic acid
730270
CC1=CC(C)=C(C)C(=C1C)S(=O)(=O)NCC(O)=O
This item is not returnable. View return policy
379250-94-9
C12H17NO4S
271.33
MFCD02333806
FAIKALCUIWGIHB-UHFFFAOYSA-N
2,3,5,6-tetramethyl-benzenesulfonylamino-acetic acid, 2,3,5,6-tetramethylphenyl sulfonyl glycine, glycine,n-2,3,5,6-tetramethylphenyl sulfonyl, 2-2,3,5,6-tetramethylbenzenesulfonamido acetic acid, 2,3,5,6-tetramethylbenzenesulfonamido acetic acid, n-2,3,5,6-tetramethylphenylsulfonyl glycine, n-2,3,5,6-tetramethylphenyl sulfonyl glycine, 2-2,3,5,6-tetramethylphenylsulfonamido acetic acid, 2,3,5,6-tetramethylbenzenesulfonamidoacetic acid, 2,3,5,6-tetramethylphenyl sulfonyl amino acetic acid
730270
CC1=CC(C)=C(C)C(=C1C)S(=O)(=O)NCC(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 166°C to 167°C
IUPAC Name 2-[(2,3,5,6-tetramethylphenyl)sulfonylamino]acetic acid
Formula Weight 271.34
Percent Purity 96%
Quantity 250 mg
Chemical Name or Material N-(2,3,5,6-Tetramethylphenylsulfonyl)glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'