missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,2,2-Triphenylacetamide, 95%

Brand:  Alfa Aesar™ H65005.03

Code : NEW

Additional Details : CAS Number : 4695-14-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402449

  • £47.81 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N,2,2-Triphenylacetamide
287.36
2127591
4695-14-1
MFCD00086997
2,2-Diphenylacetanilide

Specifications

N,2,2-Triphenylacetamide
4695-14-1
95%
MFCD00086997
2,2-Diphenylacetanilide
1g
Solid
287.36
2127591
Videos
SDS