missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3737053
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
1465-25-4
MFCD00012556
MZNYWPRCVDMOJG-UHFFFAOYSA-N
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
15106
CHEBI:53452
1465-25-4
MFCD00012556
MZNYWPRCVDMOJG-UHFFFAOYSA-N
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
15106
CHEBI:53452

N-(1-Naphthyl)ethylenediamine, dihydrochloride, >-98.0% (on dry substance), C12H16Cl2N2, CAS Number-1465-25-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Decomposition Information 200°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C12H16Cl2N2
Linear Formula C10H7NHCH2CH2NH2·2HCl
Quantity 25 g
Beilstein 12, II, 699
Sensitivity (to sulfanilamide) Passes
Merck Index 15, 6491
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot water, slightly soluble in acetone and absolute alcohol
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.174
Formula Weight 259.18
Percent Purity ≥98.0% (on dry substance)
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material N-(1-Naphthyl)ethylenediamine, dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(1-Naphthyl)ethylenediamine, dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'