missing translation for 'onlineSavingsMsg'
Learn More

N,1-Di-Boc-L-histidine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7054397
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
31687-58-8
C28H48N4O6
536.714
MFCD06804675
WBGMQHNUPJENDC-MERQFXBCSA-N
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
16211319
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
This item is not returnable. View return policy
31687-58-8
C28H48N4O6
536.714
MFCD06804675
WBGMQHNUPJENDC-MERQFXBCSA-N
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
16211319
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 160°C to 165°C (decomposition)
Formula Weight 536.7
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N,1-Di-Boc-L-histidine dicyclohexylammonium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'