missing translation for 'onlineSavingsMsg'
Learn More

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™

Catalog Number p-4509837
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
15205-27-3
C9H12NO2
166.20
MFCD04496422
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
421238
C[NH2+]CC1=CC=C2OCOC2=C1
This item is not returnable. View return policy
15205-27-3
C9H12NO2
166.20
MFCD04496422
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
421238
C[NH2+]CC1=CC=C2OCOC2=C1
Assay Percent Range 97%
Quantity 250 mg
IUPAC Name [(2H-1,3-benzodioxol-5-yl)methyl](methyl)azanium
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'