Learn More
Mystaire™ Aura™ Particulate Prefilters
For Aura™ Series Ductless Fume Hoods. An electrostatically charged fiber filter which effectively removes submicron particulates. Designed to protect the adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates.
Brand: Mystaire™ FE-3013
Code : 04
Additional Details : Weight : 14.00000kg
Description
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Prefilter | |
6 Filters | |
Electrostatically Charged Fiber | |
Prefilter |
Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood | |
Replacement Filter | |
Submicron Particulates |