missing translation for 'onlineSavingsMsg'
Learn More

Smooth Muscle Myosin, Alexa Fluor 488, clone: SMMS-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-6400-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15520237

  • £176.88 / 100µg


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

Description: This SMMS-1 monoclonal antibody reacts with both isoforms of the human smooth muscle myosin heavy chain (SM1 and SM2) structural proteins, which are components of the contractile apparatus in smooth muscle cells. Smooth muscle myosin heavy chains are expressed by myoepithelial cells and also serve as the terminal differentiation marker for smooth muscle cells. In normal breast tissue, myoepithelial cells compose the outer layer of the lumen lining. In benign and in situ breast lesions, the intact myoepithelial layer stains with SMMS-1 and the loss of this staining indicates invasive cancer. This SMMS-1 antibody has also been reported to cross-react with bovine, canine, feline, and rat smooth muscle myosin heavy chains.Applications Reported: This SMMS-1 antibody has been reported for use in microscopy, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This SMMS-1 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 30 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Myosin
SMMS-1
Alexa Fluor 488
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Microscopy
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
smooth muscle myosin heavy chains, SM1 and SM2 isoforms
IgG1, kappa
RUO
Primary
Rat
Videos

For Research Use Only.