Learn More
MYOM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556246
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CRCPIQGLVE GQSYRFRVRA ISRVGSSVPS KASELVVMGD HDAARRKTEI PFDLGNKITI STDAFEDTVT IPSPPTNVHA SEI Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.
MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.Specifications
MYOM3 | |
Polyclonal | |
Unconjugated | |
Myom3 | |
8430427K15Rik; Gm434; MYOM3; myomesin 3; Myomesin family member 3; myomesin family, member 3; myomesin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127294 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VTT5 | |
Myom3 | |
Recombinant protein corresponding to Human MYOM3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only