Learn More
MYOM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561907
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FDETRFHAGA STMPLSFGVT PYGYASRFEI HFDDKFDVSF GREGETMSLG CRVVITPEIK HFQPEIQWYR NGVPLSPSKW VQTLWSGERA TLTFSHLNKE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
MYOM1 Polyclonal antibody specifically detects MYOM1 in Human samples. It is validated for Immunohistochemistry.Specifications
MYOM1 | |
Polyclonal | |
Unconjugated | |
Myom1 | |
190 kDa connectin-associated protein; 190 kDa titin-associated protein; D430047A17Rik; EH-myomesin; MYOM1; myomesin (M-protein) 1 (190kD); myomesin 1; myomesin 1 (skelemin) 185kDa; myomesin 1, 185kDa; myomesin family member 1; Myomesin-1; skelemin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8736 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52179 | |
Myom1 | |
Recombinant protein corresponding to Human MYOM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only